CE International Trading Mesh Screen, Nylon, 2.00 mm Sieve Opening, 9 Mesh, 18" dia
Representative image only.
CE International Trading Mfr # LS450N006-#9MESH – Item # WW-59979-74
Mesh screens separate oversized particles from your sample
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNylon
  • U.S. Standard Screen Size (No.)10
  • Tyler Standard Screen Size (mesh)9
  • Sieve Opening (mm)2
  • Outside Diameter (in)18
  • DescriptionMesh Screen, Nylon, 2.00 mm sieve opening, 9 mesh
More About this Item
Screens are 18" (45.7 cm) in diameter with holes for securing in the screen assembly (sold separately under part number 59998-54).