Cascade Sciences Pure Path 100 + Kit A Thin Film Distillation System
Representative image only.
Cascade Sciences Mfr # PUREPATH100 – Item # WW-42000-17
Optimized for quick processing times
  • Two to three liter per hour throughput
  • Modular system is expandable
  • Professional installation is included in price
SPECS & DESCRIPTION
Specifications & Description
  • Condenser TypeHelical
  • Evaporation Flask Size2 L Feed Vessel
  • Power (VAC)120 / 240
  • DescriptionThin Film Distillation System
Key Features
  • Two to three liter per hour throughput
  • Modular system is expandable
  • Professional installation is included in price
More About this Item
This thin-film/wiped-film distillation system features the AG! Asahi DN100 thin film evaporator and precision glassware, Heidolph Hei-TORQUE Precision 200 overhead stirrer, Huber temperature control (CC-202C Heating Feed Vessel, Ministat 125 Cooling Condenser, CC-304B Heating Jacket) and cold traps (Minichiller 600 OLÉ glass coil condenser, TC100 cold finger), and Agilent vacuum system (DS302 Rotary Vane Pump, 8.2 CFM). With professional installation included in the price, this is a turnkey solution for any busy laboratory.
A UL/CUL listed electrical panel allows the system to run on a single 120V - 240V / 1PH / 50AMP service.