CappOrigami™ Reagent Reservoir, Polypropylene, Non-Sterile, 30 mL; 50/Pk
Representative image only.
Capp Mfr # CA40505 – Item # WW-25019-60
Innovative design minimizes reagent waste and saves valuable storage space
  • Designed to be shipped and stored as flat sheets to save space and reduce shipping costs
  • Constructed of durable and chemical-resistant polypropylene
  • Accommodates 8- and 16-channel pipettors
  • 3 year warranty
PRODUCT OPTIONS
₹8,100.00INR / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (mL)30
  • Wetted MatterialPolypropylene
  • Package Of50/Pk
  • Compatible with8- and 16-channel pipettors
  • DescriptionReagent Reservoir, Polypropylene, Non-Sterile, 30 mL; 50/Pk
  • Warranty3 year
3 Year Warranty
Key Features
  • Designed to be shipped and stored as flat sheets to save space and reduce shipping costs
  • Constructed of durable and chemical-resistant polypropylene
  • Accommodates 8- and 16-channel pipettors
  • 3 year warranty
More About this Item
CAPP® CappOrigami™ reagent reservoirs are designed to be shipped and stored as flat sheets to save space and reduce shipping costs. The polypropylene construction features a sharp V-shape that makes pipetting easier, prevents the formation of bubbles in the pipette tips, and allows you to use more accurate amounts of reagent and avoid expensive waste. The easy- to-use design features prefolded lines — simply push the sides to ''assemble''. The folds ensure that the reservoirs stay upright.