BW Technologies GasAlertMicro 5 Multigas Detector; O2/SO2/CO/H2S/LEL, Data Logger, Pump

Representative image only.
BW Technologies Mfr # M5-XWSY-R-P-D-Y-N-00 – Item # WW-10107-65
Monitors and displays up to five potential atmospheric hazards
  • Full function self-test of: sensor, battery and circuit integrity; and audible/ visual alarms
  • Compact and lightweight
  • Rugged, highly water-resistant design
  • Continuous LCD shows simultaneous gas concentrations for up to five gases
  • Integral concussion-proof enclosure for the most rugged environments
  • Integral motorized pump option for remote sampling (optional)
  • Two power options: three AAA alkaline batteries or a rechargeable battery pack
  • Provides 95dB alarm tone (typical), two bright wide-angled alarm bars and internal vibrating alarm for high-noise areas
  • Multi-language supporting English, French, German, Spanish and Portuguese
  • Four alarm levels: instantaneous Low and High alarm for all gases; TWA (time weighted average) and STEL (short term exposure limit) for toxic gases
  • Simple automatic calibration procedure: compatible with MicroDock II system
  • Records TWA, STEL and maximum exposures to gas, and displays readings on command
  • The GasAlertMicro 5 has user-selectable field options which include: confidence beep, pass code protection, latching alarm, “SAFE” display function, stealth mode, span level, and selectable ppm resolution for some gases

SPECS & DESCRIPTION

Specifications & Description

  • Gases MeasuredCO, combustibles (LEL), H2S, O2, SO2
  • Min CO Range (ppm)0
  • Max CO Range (ppm)999
  • Min Oxygen Range (%)0
  • Max Oxygen Range (%)30
  • Min Hydrogen Sulfide Range (ppm)0
  • Max Hydrogen Sulfide Range (ppm)500
  • Min Combustible Range (% LEL)0
  • Max Combustible Range (% LEL)100
  • AlarmAudible (95 dB), visual, and vibrating
  • Battery TypeRechargable battery
  • Battery Life15
  • Height (in)5 3/4
  • Width (in)2 7/8
  • Depth (in)1 1/2
  • Height (cm)14.6
  • Width (cm)7.3
  • Depth (cm)3.8
  • DescriptionMultigas Detector; O2/SO2/CO/H2S/LEL, Data Logger, Pump
Two year warrantyConformite EuropeenneCanadian Standards AssociationIntrinsically SafeIP 65IP 67

Key Features

  • Full function self-test of: sensor, battery and circuit integrity; and audible/ visual alarms
  • Compact and lightweight
  • Rugged, highly water-resistant design
  • Continuous LCD shows simultaneous gas concentrations for up to five gases
  • Integral concussion-proof enclosure for the most rugged environments
  • Integral motorized pump option for remote sampling (optional)
  • Two power options: three AAA alkaline batteries or a rechargeable battery pack
  • Provides 95dB alarm tone (typical), two bright wide-angled alarm bars and internal vibrating alarm for high-noise areas
  • Multi-language supporting English, French, German, Spanish and Portuguese
  • Four alarm levels: instantaneous Low and High alarm for all gases; TWA (time weighted average) and STEL (short term exposure limit) for toxic gases
  • Simple automatic calibration procedure: compatible with MicroDock II system
  • Records TWA, STEL and maximum exposures to gas, and displays readings on command
  • The GasAlertMicro 5 has user-selectable field options which include: confidence beep, pass code protection, latching alarm, “SAFE” display function, stealth mode, span level, and selectable ppm resolution for some gases

More About this Item

GasAlertMicro 5 portable gas detector simultaneously monitors and displays up to five potential atmospheric hazards including oxygen, combustible gases and a range of toxic gases. Compact, lightweight and water-resistant. Audible, visual and vibrator alarms activate in the event of a low, high, TWA or STEL alarm condition. An optional motorized pump is available.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat