Burkle 5379-0009 Disposable sampling spatula, PE, FDA compliant, white; 150 mm
Representative image only.
Burkle Mfr # 5379-0009 – Item # WW-06042-86
Ecologically sustainable labware constructed of renewable raw materials
  • Packaged individually and suitable for food-grade applications
  • Produced and packaged according to ISO Class 7
  • EU foodstuffs and FDA compliant
  • 15 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)10 2/5
  • Overall Length (cm)26.3
  • MaterialPolyethylene
  • SterileNo
  • DescriptionDisposable sampling spatula, PE, FDA compliant, white; 150 mm
  • Warranty15 month
15 Month Warranty
Key Features
  • Packaged individually and suitable for food-grade applications
  • Produced and packaged according to ISO Class 7
  • EU foodstuffs and FDA compliant
  • 15 month warranty
More About this Item
The disposable sampling spatulas are long and V-shaped with a stable handle and sharp blade. This makes it possible to penetrate directly into containers such as paper or plastic sacks. These spatulas are ideal for sampling powders, granulates, and pastes. The total length of these spatulas is 26.3 cm with an insertion depth of 15 cm. These are available nonsterilized or presterilized via gamma irradiation.