This item is now discontinued. We recommend the following as suggested replacements: WW-06101-31
Burkle 0340-0100 Reagent Bottle, Narrow Neck, HDPE, 32 mm Cap; 100 mL
Representative image only.
Burkle Mfr # 0340-0100 – Item # WW-06040-27
Accessories for the Compact Fluid Sampling Kits
  • Eliminate cross-contamination while taking samples
  • Durable components suitable for use with viscous oils
  • Quick and comfortable sampling with hand-operated vacuum-piston pump
  • 15 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHDPE
  • Bottle Type100-mL HDPE
  • Cap Configuration32 mm
  • DescriptionBurkle, Inc. 0340-0100 Reagent bottle, narrow neck, HDPE, 32 mm cap; 100 mL
  • Warranty15 month
15 Month Warranty
Key Features
  • Eliminate cross-contamination while taking samples
  • Durable components suitable for use with viscous oils
  • Quick and comfortable sampling with hand-operated vacuum-piston pump
  • 15 month warranty
More About this Item
The compact fluid sampling systems greatly reduce the possibility of cross-contamination as the liquid only comes into contact with the suction hose and sample bottle. After the sample is taken, the bottles are sealed, and the suction tubing can quickly and easily be replaced. The small-diameter 8-mm hose can reach narrow, hard-to-access sampling areas. Both models can be used with the PTFE adapter for glass thread GL45 for enhanced chemical resistance and use with other sampling bottles.
Kit 06040-94 is for general sampling from barrels, canisters, tanks, and more, as the wetted materials are polyethylene. This kit can be with the 06040-27 HDPE sampling bottle and polyethylene suction hose 06044-36 (10 m roll) or 06044-37 (100 m roll).
Kit 06040-95 is for ultrapure sampling and ideal when maximum purity or chemical resistance is required. This pump can be used with the PTFE suction hose 06044-35 (5 m roll).
Sampling kits sold separately.