Bud Industries TR-6101 Enclosure for DIN-Mount Controllers, Medium Size
Representative image only.
Bud Industries TR-6101 Enclosure for DIN-Mount Controllers, Medium Size
Bud Industries | Mfr: TR-6101| Item: 9450318
Convert your DIN controllers into a portable desktop instrument
₹85,500.00INR / Each
₹85,500.00INR / Each
Buy It Together
Current Item
Bud Industries TR-6101 Enclosure for DIN-Mount Controllers, Medium Size
Bud Industries TR-6101 Enclosure for DIN-Mount Controllers, Medium Size
9450318
₹85,500.00INR / Each
+ =
Total Price 86130.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Corner gussets provides additional strength
  • Carrying handle may be used as tilting bail
  • 3 month warranty
More About this Item
The versitality makes it ideal to use as a portable enclosure, a desk top instrument case, or directly into a larger system. Front panel is contructed of 0.09" aluminum and can be easily stamped out to fit controllers and panel meters from 1/16 to 1/2 DIN sizes. Top and bottom panels are constructed from 0.05" aluminum and are easily removed for access to the dust-free interior. Enclosure is finished with a royal blue texture with a white frame. Enclosure comes completely assembled with all hardware included.
3 Month Warranty
Specifications
  • MaterialAluminum
  • Outside Width (in)8.31
  • Outside Height (in)5.25
  • Outside Depth (in)13.25
  • DescriptionEnclosure for DIN-Mount Controllers, Medium Size
  • Warranty3 Months