Bud Industries NF 6612 NEMA 4X Fiberglass Box with Screw Cover and Mounting Brackets
Representative image only.
Bud Industries NF 6612 NEMA 4X Fiberglass Box with Screw Cover and Mounting Brackets
Bud Industries | Mfr: NF-6612| Item: 9450184
Fiberglass-reinforced plastic provides outstanding protection against corrosion
₹21,200.00INR / Each
₹21,200.00INR / Each
Product Description
Key Features
  • Watertight, chemical resistant, durable, and impervious to dents
  • Stainless steel hinge, screws, and wall mounting brackets
  • Includes powder-coated 14-gauge steel removable inner panel
  • Light gray finish
  • NF models have a stainless steel screw-type cover; NFL models have a stainless steel latch-type cover
  • Designed to meet NEMA 1, 2, 4, 4X, 12, 13, and IP65 specifications
  • UL listed and tested to UL508-4X requirements, UL File E194432
  • 1 year warranty
1 Year WarrantyIP 65
Specifications
  • MaterialFiberglass
  • Outside Width (in)9.9
  • Outside Height (in)11.78
  • Outside Depth (in)6.8
  • DescriptionNEMA 4X Fiberglass Box with Screw Cover and Mounting Brackets