Buck Scientific GRAMS/AI (version 7.0) Infrared Analysis Software for Windows
Representative image only.
Buck Scientific Mfr # GRAMS/AI – Item # WW-83001-27
Touch screen display lets you view spectra and run survey scans without a PC Display measurement values as ratios in either percent transmission or absorbance Single beam optical design provides dual beam capabilities Nitrogen purge not required—eliminate costly gases Connects easily to a computer or recorder
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Description(version 7.0) Infrared Analysis Software for Windows
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Simply stated, this is the easiest IR spectrophotometer to operate and maintain! Advanced self-help menus guide you every step of the way. Analysis requires no expensive nitrogen purge for moisture or CO2 prior to use. Single-button controls set all instrument parameters for operation and rapid data collection. High energy optical design and a sensitive DTGS detector provides excellent resolution over the full infrared range. Maximum performance is achieved with a fast scanning mechanism that is completely microprocessor driven—you get dual-beam capabilities with just a single beam optical bed. Display simultaneously indicates wavelength, transmittance or absorbance, scan type, scan time, and response time in seconds. Easy Scan software (included) easily accomm