BTO Bleed and Temperature Optimized Septa, Shimadzu; 50/Pk
Representative image only.
BTO Mfr # 298735 – Item # WW-01999-14
Optimized for the most demanding GC and GC/MS applications
  • Formulated to extend low-bleed and outstanding mechanical properties
  • 1 month warranty
PRODUCT OPTIONS
₹25,300.00INR / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • Description(Bleed/Temp Optimized) Septa, Shimadzu; 50/Pk
  • Warranty1 month
1 Month Warranty
Key Features
  • Formulated to extend low-bleed and outstanding mechanical properties
  • 1 month warranty
More About this Item
BTO® (Bleed and Temperature Optimized) are designed for low bleed and temperatures up to 400°C (752 °F). Septa changes are easy—septa will not stick to your port.