BriskHeat CPBIH101012LGT Grounded Fiberglass Insulated Heat Tape, 12' x 1" , 600 Watts, 115 V

Representative image only.
BriskHeat Mfr # CPBIH101012LGT – Item # WW-03121-60
Safe for heating metal vessels
  • Heat to 482F (250C)
  • NOTE: Temperature range has been reduced to meet safty standards
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Max Temperature (° F)482
  • Max Temperature (° C)250
  • Plug TypeNo plug
  • Power (VAC)115
  • Power (watts)600
  • Width (in)1
  • Length (ft)12
  • DescriptionGrounded Fiberglass Insulated Heat Tape, 12' x 1\" , 600 Watts, 115 V
  • Warranty1 year
1 Year Warranty

Key Features

  • Heat to 482F (250C)
  • NOTE: Temperature range has been reduced to meet safty standards
  • 1 year warranty

More About this Item

Wind heating tape around glass, ceramic, or metal vessels—grounding prevents shock hazard. Temperature-resistant fiberglass sleeves encase heating element; ground wire at core. The maximum surface loading is 4 watts/sq in. Leads are 31/2" long with electrical connector at one end. Tapes include ties at both ends.

WARNING: Heating tape should not be overlapped and require the use of a temperature or power controller sold separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat