This item is now discontinued. We recommend the following as suggested replacements: WW-18100-05
Branson Industrial strength ultrasonic cleaning solution, 1 quart bottle
Representative image only.
Branson Mfr # 000-955-114 – Item # WW-08801-21
Safely remove grease and oils
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (gallons)0.25
  • Volume (Liters)1
  • Dilution8 to 10%, 11 to 12 pH
  • DescriptionIndustrial strength ultrasonic cleaning solution, 1 quart bottle
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Removes grease, oils, and particulates from automotive, aircraft, and similar mechanical components. This cleaner cleans shop oils and light carbon deposits from valves fittings, and similar components.