BRANDplates® cellGrade™ Sterile Microplate, 1536-Well, PS, Black, 10 µL, Standard F-Bottom; 50/PK
Representative image only.
Mfr # 782002 – Item # WW-07903-73
Minimizes cross-contamination and ensures maximize results
  • cellGrade™ standard cell culture treatment
  • cellGrade™ plus enhanced surface for more fastidious cells, or reduced serum culture
  • cellGrade™ preminum poly-D lysine equivalent for serum-free culture, or the most fastidious cells
  • inertGrade for suspension cultivation to inhibit differentiation of stem cells
  • Sterility Assurance Level (SAL) of 10-6
  • Ideal for the cultivation of adherent cell lines with a hydrophilic surface
  • Volume: 10 µL
  • Standard F-Bottom
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells1536
  • Well Volume (µL)10
  • Well ShapeStandard F-Bottom
  • Surface Treatment cellGrade™
  • Sterile Sterile
  • Plate ColorBlack
  • MaterialPolystyrene
  • DescriptionBrandTech 782002 BRANDplates® cellGrade™, Sterile Microplate, 1536-Well, PS, Black, 10 µL, Standard F-Bottom; 50/PK
Key Features
  • cellGrade™ standard cell culture treatment
  • cellGrade™ plus enhanced surface for more fastidious cells, or reduced serum culture
  • cellGrade™ preminum poly-D lysine equivalent for serum-free culture, or the most fastidious cells
  • inertGrade for suspension cultivation to inhibit differentiation of stem cells
  • Sterility Assurance Level (SAL) of 10-6
  • Ideal for the cultivation of adherent cell lines with a hydrophilic surface
  • Volume: 10 µL
  • Standard F-Bottom
More About this Item
These cell culture microplates come in a variety of surface treatments ideal for use in the culture of a wide range of cell types. The plates are sterilized by ß-radiation or ethylene oxide, are DNA-, DNase-, RNase-free, and are packaged individually wrapped.
The polystyrene microplates are designed to lessen cross contamination, ensure outstanding recovery, and offer many options with color and number of wells. Three different bottom shapes are available: round (U) with no edges, flat (F) with a flat bottom, and flat with curved edges for enhanced mixing (C) bottom. For easy identification of the surface type and well location, the 96-well standard format plates feature an orange color-coded embossed alphanumeric lettering for easy identification of wells.
With a chimney well type of design, each well is separated from each other to reduce the risk of cross contamination. The 384-well microplates feature rounded corners which allow optimal uniform meniscus formation.
Convenient accessories for use with the microplates, such as lids, rubber rollers, and sealing films are offered.