MTC Bio BowTie™ Biohazard Bin with Lid and Foot Pedal, 12 to 14 Gallon, PP

Representative image only.
MTC Bio Mfr # A8000B – Item # WW-14003-02
Hands-free foot pedal operated lid keeps contamination to a minimum
  • Compatible with BowTie™ biohazard bags
  • Clearly marked with word "Biohazard" and biohazard symbol

₹13,100.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (Liters)52
  • Capacity (Gal)14
  • MaterialPolypropylene (PP)
  • ColorRed
  • Compatible WithBowTie™ biohazard bags
  • Length (in)14
  • Height (in)19
  • Width (in)10
  • Width (cm)25
  • Height (cm)48
  • Length (cm)35
  • DescriptionBowTie™ Biohazard Bin with Lid and Foot Pedal, 12 to 14 Gallon, PP

Key Features

  • Compatible with BowTie™ biohazard bags
  • Clearly marked with word "Biohazard" and biohazard symbol

More About this Item

Touchless lid offers quick disposal of hazardous waste. Bin measures 14"L x 10"W x 19"H (35 x 25 x 48 cm) and works perfectly with BowTie biohazard bags.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat