This item is now discontinued. We recommend the following as suggested replacements: WW-06286-01
Frozen Probe Cylindrical Sampler, 24 mL capacity, 9.5"L
Representative image only.
Item # WW-33687-40
Perfect for sampling frozen materials
  • Constructed of stainless steel
  • Can be autoclaved
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)9.5
  • DescriptionFrozen Probe Cylindrical Sampler, 24 mL capacity, 9.5\"L
  • Warranty1 Year
1 Year Warranty
Key Features
  • Constructed of stainless steel
  • Can be autoclaved
  • 1 year warranty
More About this Item
Cylindrical probe makes collecting samples from frozen blocks easy. Use the probe with an electric drill—simply start the drill and begin to insert the probe inside the frozen material. Once inserted, the sample is collected. To release the sample, detach probe from drill and push out the sample with the expelling rod. Probe comes complete with standard drill attachment and expelling rod.