BioBundle Fermentation Polarization Dissolved Oxygen Sensor; 2-Pin Low Drift
Representative image only.
BioBundle Mfr # Z110001020 – Item # WW-29207-46
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFermentation Polarization Dissolved Oxygen Sensor; 2-Pin Low Drift
More About this Item
These polarization DO sensors are autoclavable. These sensors have built-in battery that keeps the sensor charged, eliminate 6 to 10 hours waiting for calibration, and are ready to go whenever the operator wants.