Marsh Bellofram 961-074-000 Pressure Transmitter, I to P;3-27 PSI, 4-20 mA
Representative image only.
Marsh Bellofram Mfr # 961-074-000 – Item # WW-68826-02
High-flow capacity up to 12 scfm
  • 4 to 20 mA Input
  • Mounts in any angle
  • Small size and lightweight
  • External zero and span adjustments
  • 18 month warranty
PRODUCT OPTIONS
₹54,700.00INR / Each
₹27,000.00 - ₹33,700.00INR / Each
for an additional
₹27,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Min Exhaust Flow Rate (SCFM)2
  • Forward Flow Capacity (SCFM)24
  • Forward Flow Capacity (LPM)677
  • Min Operating Temperature (° F)-20
  • Max Operating Temperature (° F)160
  • Min Operating Temperature (° C)-28
  • Max Operating Temperature (° C)71
  • Control Range (low) (PSI)3
  • Control Range (high) (PSI)27
  • Process Connection NPT(F) (in)1/4
  • Electrical Connections¼\" NPT(F)
  • DescriptionPressure Transmitter, I to P;3-27 PSI, 4-20 mA
  • Warranty18 months
18 Month Warranty
Key Features
  • 4 to 20 mA Input
  • Mounts in any angle
  • Small size and lightweight
  • External zero and span adjustments
  • 18 month warranty
More About this Item
These transducers are an electro-pneumatic device that reduces supply pressure to a regulated output pressure directly proportional to an electrical input signal. These transducers are also field-reversible by reversing the polarity of the signal leads and recalibrating. After calibration, the transducer will then provide a minimum of it''s full rated output pressure upon input signal failure. These units are typically used in HVAC-R systems, petrochemical processing, and slurry processing.