Bel-Art F18385-0000 Polystyrene Antivibration Damping Mount Base, 12.5 x 15.25 x 4.5"
Representative image only.
Bel-Art Mfr # F18385-0000 – Item # WW-01019-40
Reduce vibration disturbances caused by nearby equipment
₹149,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)4 1/2
  • Width (in)15 1/4
  • Depth (in)12 1/2
  • DescriptionPolystyrene Antivibration Damping Mount Base, 12.5 x 15.25 x 4.5\"
More About this Item
The economical polystryene mount offers an easy-to-clean neoprene work surface which reduces spillage; nonslip rubber feet provide enhanced stability.