Bausch & Lomb Measuring Scale, Inch, 0.75” in 0.005” Increments
Representative image only.
Bausch & Lomb Mfr # 81-34-37 – Item # WW-03882-12
Improve inspection measurements
  • Glass measuring scales are compatible with measuring magnifiers
  • 1 year warranty
PRODUCT OPTIONS
₹12,200.00INR / Each
Include Measuring Magnifier…
SPECS & DESCRIPTION
Specifications & Description
  • MagnificationNone
  • Lens0.75" Scale with 0.005" increments
  • Mounting TypeCompatible with Hastings Measuring Magnifiers
  • DescriptionMeasuring Scale, Inch, 0.75” in 0.005” Increments
  • Warranty1 Year
1 Year Warranty
Key Features
  • Glass measuring scales are compatible with measuring magnifiers
  • 1 year warranty
More About this Item
Use these measuring scales with the Hastings measuring magnifiers. Scales simply screw onto the bottom of each magnifier lens.
Measuring magnifiers 03882-10 and -50 are sold separately.