Bausch & Lomb Coddington Magnifier, 20X, 0.49" Diameter
Representative image only.
Bausch & Lomb Mfr # 81-61-41 – Item # WW-03883-14
Provide good definition and a wide field of view
  • Durable nickel-plated case protects the lens
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification20x
  • Viewing Area0.49\" (12.4 mm) diameter
  • LensSingle
  • Focal Length0.5\" (12.7 mm)
  • BatteryNone
  • DescriptionCoddington Magnifier, 20x Magnification
  • Warranty1 Year
1 Year Warranty
Key Features
  • Durable nickel-plated case protects the lens
  • 1 year warranty
More About this Item
These high-performance Coddington magnifiers provide sharp, crisp images. Correction is achieved through the use of a single thick lens with a central groove diaphragm. Swing-away, nickel-plated case protects the lens and acts as a handle.