HDPE Covers for Basins 06019-10, -20; 12/PK
Representative image only.
Item # WW-06019-01
₹21,900.00INR / Pkg of 12
SPECS & DESCRIPTION
Specifications & Description
  • ColorGray
  • Cap MaterialHigh-Density Polyethylene (HDPE)
  • DescriptionHDPE Covers for Basins 06019-10, -20; 12/PK
NSF international certified
More About this Item
Basin cover is made of high-density polyethylene (HDPE). Temperature range is -40 to 180°F (-40 to 83°C). Contains no BPAs. Basins sold separately.