B&K Precision 601B Handheld Battery Capacity Analyzer; 6 and 12 V SLA
Representative image only.
B&K Precision Mfr # 601B – Item # WW-20025-99
Test and maintain sealed lead acid (SLA) batteries
  • Measure battery voltage and capacity
  • Powered by the battery under test—no need for an external power supply
  • Rugged housing with durable Kelvin clamps
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • AC Max (Volts)6
  • DescriptionHandheld Battery Capacity Analyzer; 6 and 12 V SLA
  • Warranty1 Year
1 Year Warranty
Key Features
  • Measure battery voltage and capacity
  • Powered by the battery under test—no need for an external power supply
  • Rugged housing with durable Kelvin clamps
  • 1 year warranty
More About this Item
These lightweight, handheld battery capacity analyzers provide a quick, accurate test of percent capacity and voltage in 12 V lead acid batteries. Easily portable and ideal for field and shop service, maintenance applications with automotive fleets, communication equipment, UPS manufacturing and maintenance, and backup lighting systems.