Autoclave Tape, 12 mm Wide, 8 X 50 M Roll
Representative image only.
Item # WW-94702-17
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)1/2
  • Width (cm)12
  • DescriptionAutoclave Tape, 12 mm Wide, 8 X 50 M Roll
More About this Item
Autoclave Tapes - Ideal for Autoclave validation: The process indicator stripes turn white to dark brown / black when exposed to either steam sterilization conditions in a 121 degC gravity sterilizer or a 132-134 degC pre-vacuum sterilizer. Also available masking tape (without indicator). Meets 510 K Notified compliance. ETO Tape: The process indicator stripes turn brown to green after exposure to an EO gas sterilization process operating at 600 mg/L EO gas, 50% ±10% relative humidity, 54.4degC for 45 minutes or longer.