Ashcroft Pressure Switch, 400 PSI, 1/4" NPT(F); Single Set PointPressure Switch, 100" WC, 1/4" NPT(F)
Representative image only.
Ashcroft Mfr # LPAN4HB25400# – Item # WW-68360-09
Designed for general-purpose to highly caustic applications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Process Connection1/4\" NPT(F)
  • DescriptionPressure Switch, 400 PSI, 1/4\" NPT(F); Single Set PointPressure Switch, 100\" WC, 1/4\" NPT(F)
  • Warranty1 year
1 Year WarrantyConformite EuropeenneCanadian Standards AssociationFM ApprovedIP 66 RatedUL Listed.
Key Features
  • 1 year warranty
More About this Item
Watertight, general purpose pressure switch is designed for use in applications requiring high reliability, safety and accuracy. Its straight forward design allows for easy field installation and calibration. Single setpoint with fixed or adjustable deadband or two independent set points with fixed deadband units are available. Aluminum housing is epoxy coated for durability and long life.
Switches have a wide range of uses — choose from inches WC to high pressure in gauge or differential ranges. Applications include pulp and paper mills, pharmaceutical manufacturing, dairies, and breweries.