Ashcroft Pressure Switch, 100" WC, 1/4" NPT(F); Two Set Points
Representative image only.
Ashcroft Mfr # GPDN4GGB25100IW – Item # WW-68360-38
316L stainless steel housing allows use in highly caustic applications
  • Advanced switches provide single or adjustable set points
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Process Connection1/4\" NPT(F)
  • DescriptionPressure Switch, 100\" WC, 1/4\" NPT(F); Two Set Points
  • Warranty1 year
1 Year WarrantyConformite EuropeenneCanadian Standards AssociationIP 65UL Listed.
Key Features
  • Advanced switches provide single or adjustable set points
  • 1 year warranty
More About this Item
Watertight general purpose pressure switch is designed for use in tough applications requiring a 316L SS enclosure. The simple design allows for easy field installation and calibration, while proven technology ensures a highly reliable, safe and accurate switch. Choose from switches that feature single setpoint with fixed or adjustable deadband or two independent set points with fixed deadband to best suit your application.
Designed for use in chemical and petrochical manufacturing, water and wastewater plants, steel mills, or other tough environments. Pressure switches allow for use in a wide variety of applications with their large range of pressure from inches WC to high pressure in gauge and differential.