Ashcroft 4 1/2 Low-pressure Bellows Gauge, 0 To 15 Inwc, 1/4 NPT
Representative image only.
Ashcroft Mfr # 451188SS 04L 15IW/9ZI – Item # WW-68427-84
The Ashcroft® 1188 bellows gauge is used when an ASME style low pressure gauge is required
  • A variety of wetted parts compatable with many process medias
  • Ideal product solution for many installations
  • Overpressure capability up to 5x full scale range standard
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±2-1-2% full-scale (ANSI/ASME Grade A)
  • Dial Size (in)4.5
  • Dial Size (mm)114
  • Min Pressure (in WC)0
  • Max Pressure (in WC)15
  • Min Temperature (° F)-4
  • Max Temperature (° F)185
  • Min Temperature (° C)-20
  • Max Temperature (° C)85
  • Wetted Materials316 SS
  • CasePhenolic
  • Lens Materialpolycarbonate
  • Process Connection1/4\" NPT(M)
  • Unit Of Measurement\" WC, oz/in²
  • DisplayAnalog dial
  • Media CompatibilityGases and liquids compatible with wetted materials
  • Description4 1/2 Low-pressure Bellows Gauge, 0 To 15 Inwc, 1/4 NPT
  • Warranty1 year
1 Year Warranty
Key Features
  • A variety of wetted parts compatable with many process medias
  • Ideal product solution for many installations
  • Overpressure capability up to 5x full scale range standard
  • 1 year warranty
More About this Item
Its highly sensitive system and standard product features are available to meet many process applications. Available with diaphragm seals, this is an ideal product instrument solution for many installation points. With ranges as low as 10 inches of water vacuum, a variety of wetted parts and case materials and solid front case designs, these low pressure gauges are generally used in applications such as oil & gas, chemical/petrochemical plants, water/wastewater pressure control, and equipment skids.