Arioso Power Integrate Water Purification System with UV, TOC & Ultrafilter, 35 L/hr; 120 VAC

Representative image only.
Arioso Mfr # AP3-I-TOC-120 – Item # WW-67000-49
Single unit delivers pure or ultrapure water with one touch
  • Touch screen and plain language menus make setup easy
  • Dedicated pure and ultrapure water dispense icons on main screen simplify operation
  • Open front door to quickly change treatment cartridges
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Water Purification MethodReverse osmosis and ultrapure treatment
  • Max Production (L/hour)35
  • Inlet Port6-mm OD tubing
  • With UV Lamp254/185 nm
  • With Ultrafilter5000 MW
  • TOC MonitorYes, 0 to 5 ppb
  • Feed Water Requirements (Source)Tap or ground water
  • Type I Water (Resistivity)Ultrapure Water: up to 18.3 MΩ•cm
  • Feed Water Requirements (Conductivity)0 to 999 µS/cm
  • Feed Water Requirements (Temperature)0 to 100°C (32 to 212°F)
  • Type II Water (Conductivity)Pure Water: 0.2 to 250.0 µS/cm
  • Min Operating Pressure (PSI)14.5
  • Max Operating Pressure (PSI)72.5
  • Min Operating Pressure (bar)1
  • Max Operating Pressure (bar)5
  • Power (VAC)120
  • Height (in)18.75
  • Depth (in)19.25
  • Width (in)12.625
  • Height (cm)47.5
  • Depth (cm)49
  • Width (cm)32
  • DescriptionWater Purification System with UV, TOC & Ultrafilter, 35 L/hr; 120 VAC
  • Warranty1 Year
1 Year Warranty

Includes

TOC models include meter, flow controller, and tubing.

Key Features

  • Touch screen and plain language menus make setup easy
  • Dedicated pure and ultrapure water dispense icons on main screen simplify operation
  • Open front door to quickly change treatment cartridges
  • 1 year warranty

More About this Item

This combination reverse osmosis (RO) and ultrapure (UP) treatment water purification system provides a complete solution to your laboratory-grade water requirements. Pure water, produced by the RO component, is suitable for general laboratory applications, such as reagent and media preparation, glassware rinsing, and autoclaving. Ultrapure water, produced when both RO and UP components are used in conjunction, is necessary for sensitive analytical applications, such as high-performance liquid chromatography (HPLC), gas chromatography (GC), mass spectroscopy (MS) and other critical uses.

Compact unit fits on the bench top near your workspace for access at point of use. Convenient tap water supply feed works in any lab.

Integrate Models provide the best water quality, combining UV disinfection and the 5000 MWCO ultrafilter. The 25 and 35 L/hr units have the option of TOC indication.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat