ResinTech VP-17-3680-HN VP Series - Oxygen Reduction Filter Cartridge, Hose Nipple
Representative image only.
ResinTech Mfr # VP-17-3680-HN – Item # WW-99284-90
Reduce operating costs while maintaining peak performance
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Made in the USA!
  • Oxygen removal Hose barb, 3/8"
  • 1 year warranty
PRODUCT OPTIONS
₹41,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MediaOxygen Removal
  • Capacity1980 ppm
  • Connection2 x 3/8\" Hose Barb
  • Compatible withARIES, Unix, Barnstead (8903), Vaponics (OXN-1)
  • Max Inlet Pressure (PSI)30
  • Max Flow Rate (LPM)3.8
  • Max Flow Rate (GPH)60
  • Diameter (in)3.5
  • Length (in)17.8
  • DescriptionFilter Cartridge, Oxygen Removal Resin, 3/8\" Hose Barb
  • Warranty1 year
1 Year Warranty
Key Features
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Made in the USA!
  • Oxygen removal Hose barb, 3/8"
  • 1 year warranty
More About this Item
Use these filtration cartridges in your ResinTech, UNIX, Barnstead® PCS® and Vaponics® water purification systems or design a system using wall-mounting brackets and extension tubing (order separately). Permanent joint welds create a hermetic seal and improve longevity in any installation. Nonpigmented ABS plastic housing is durable and produces high-purity laboratory-grade water. Choose between the standard ARIES connection type and the versatile 3/8" hose nipple, both of which are available in a variety of filter media.