ResinTech CLiR 5200 Ultrapure Lab Water System (Standard 110v)

Representative image only.
ResinTech Mfr # CLS-5200-S-1 – Item # WW-99285-35
Obtain Type I, 18.2 MΩ water—on demand!
  • Meets ASTM Type I water specifications
  • Self Servicable! No tools required
  • Quick change consumables
  • Compact, flexible configurations save counter space
  • Built-in timer prevents excess wear
  • 2.5 LPM flowrate
  • Built-in leak detection
  • Intuitive user interface
  • Low-cost, long-life consumables
  • Lowest cost-of-ownership
  • Made in the USA!
  • 1 year warranty

PRODUCT OPTIONS

₹2,196,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • ASTM Water GradeType I
  • Max Flow Rate (L/min)2.5
  • Inlet Port3/8\" OD tube
  • With UV LampYes
  • With UltrafilterNo
  • Resistivity Threshold18.2 MΩ-cm
  • Conductivity Threshold0.056 µS/cm
  • TOC Threshold< 5 ppb
  • Feed Water Requirements (Source)RO/DI/Distilled
  • Feed Water Requirements (Conductivity)<20 µS/cm
  • Feed Water Requirements (TOC)<50 ppb
  • Type I Water (Bacteria)<1 CFU/mL
  • Feed Water Requirements (pH range)5 to 8
  • Feed Water Requirements (pressure)20 psi (1.3 bar) min, 60 psi (4.1 bar) max
  • Feed Water Requirements (Temperature)100°F (37°C) max
  • Power (VAC)120
  • DescriptionCitation Benchtop System with UV Oxidation Lamp, 120V/60Hz
  • Warranty1 year
1 Year Warranty

Key Features

  • Meets ASTM Type I water specifications
  • Self Servicable! No tools required
  • Quick change consumables
  • Compact, flexible configurations save counter space
  • Built-in timer prevents excess wear
  • 2.5 LPM flowrate
  • Built-in leak detection
  • Intuitive user interface
  • Low-cost, long-life consumables
  • Lowest cost-of-ownership
  • Made in the USA!
  • 1 year warranty

More About this Item

CLiR high-purity water systems features continual recirculation to ensures constant ultra-pure water for use in your lab. Choose from systems offering up to 4.0 liters of 18.2 megohm water per minute. Standard features include a built-in pressure regulator, high-capacity cartridges for deionizing the water, and a POU capsule capsule filter for removal of particulates and unwanted bacteria from the water as it is being dispensed through the outlet valve.

Multiple sensors continuously monitor the system and final water quality to ensure trouble-free operation. A digital resistivity monitor with alarm LED set points provides system feedback performance. The large volume cartridges require less change-outs, offering you lower operating costs. [Note: The feed water quality will determine the necessary pre-treatment. For optimum performance, the water system may require reverse osmosis (99284-51), distillation, or deionizing pretreatment.]

Small footprints and flexible mounting options (counter, wall, and under-sink) allow you to maximize valuble counter space. Consumables are simple to replace and require no tools (Cartridges sold separately.). Dispensing gun and secondary auxiliary side port are available upon request. Additional options are available for special applications to include 185/254 nm UV, and or ultrafiltration (UF).
Some CLiR system have been pre-configured into four application specific models with benchtop or remote dispesing options:
For Basic chemistry, academia, IC, and buffers: CLiR 5100 (models 99285-31 and -32)
For HPLC, GC-MS, and trace organics: CLiR 5200 (models 99285-35 and -36 with UV)
For Life science, cell culture, and microbiology: CLiR 5300 (models 99285-33 and -34 with UF)
For DNA Sequencing, PCR, and electrophoresis: CLiR 5400 (models 99285-37 and -38 with UV and UF)

A complete package requires a system, conditioning cartridge, polishing cartridge, and final filter.
1. Choose CLiR 5000 base system
2. Choose conditioning cartridge 99285-45 for RO water feed, or 99285-46 for SDI/tap water feed
3. Choose polishing cartridge 99285-43 for standard results, or 99285-44 for additional organics removal
4. Choose final filter 99285-27 for up to 0.2µm pariculate removal, or 99285-47 to also include endotoxin removal

Options (must be ordered with unit, factory installation required):
Item 99285-39 - adds in flow totalizer function, viewable on display
Item 99285-40 - adds in a direct feed port
Item 99285-41 - adds in a recirculating dispensing gun (for units without a remote dispense function)
Item 99285-42 - adds in a wall mounting bracket to clear up bench space"

Ideal for reagent grade solution make-up, ion chromatography, instrument feed, hydrogen generator, general lab water purification, clincal diagnostic, mass spectroscopy, microbiology, trace metals, and HPLC.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat