Argos Technologies Trifill Trifill Multichannel Pipette Reservoir, 10/package

Representative image only.
Item # WW-25200-71
Innovative design incorporates three types of reagent reservoir in one unit
  • Graduated single reagent reservoir with 50 mL maximum volume
  • Multichannel pipetting reservoir with eight separate 5 mL reagent channels
  • Multichannel pipetting reservoir with twelve separate 5 mL reagent channels
  • Individual compartments of the multichannel reservoirs are numbered for reagent identity
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • SterileNo
  • Max Volume (µL)5000
  • Min Volume (µL)0
  • Min Volume (mL)0
  • Max Volume (mL)5
  • Number Of Channels1
  • Wetted MatterialPolypropylene
  • DescriptionTrifill Multichannel Pipette Reservoir, 10/package
  • Warranty1 year
1 Year WarrantyConformite Europeenne

Key Features

  • Graduated single reagent reservoir with 50 mL maximum volume
  • Multichannel pipetting reservoir with eight separate 5 mL reagent channels
  • Multichannel pipetting reservoir with twelve separate 5 mL reagent channels
  • Individual compartments of the multichannel reservoirs are numbered for reagent identity
  • 1 year warranty

More About this Item

Trifill is manufactured of polypropylene and can be cleaned or steam sterilized if necessary. Reservoirs are individually wrapped in packs of ten.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat