Argos Technologies PolarSafe® Ice Bucket with Lid, 2.5 L, Polystyrene, Black; Each
Representative image only.
Argos Technologies Item # WW-04396-94
Perfectly suited for use with ice, dry ice, and slurry ice
  • Lightweight and more economical than solid plastic models
  • Provide excellent insulating properties to help keep temperature sensitive samples safely chilled
  • Designed for steady and sturdy benchtop use and will not leave condensation
  • Snug fitting lids included to minimize evaporation and maximize cooling
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (L)2.5
  • ColorBlack
  • MaterialPolystyrene
  • DescriptionIce Bucket with Lid, 2.5 L, Polystyrene, Black; Each
Key Features
  • Lightweight and more economical than solid plastic models
  • Provide excellent insulating properties to help keep temperature sensitive samples safely chilled
  • Designed for steady and sturdy benchtop use and will not leave condensation
  • Snug fitting lids included to minimize evaporation and maximize cooling
More About this Item
Buckets and pans are cool containers made of expanded polystyrene that are an economical alternative to polyurethane or other solid-plastic versions. Built in handles and lips make them easy to carry.