Argos Technologies Pipetting Work Station without brackets (2/pk)
Representative image only.
Argos Technologies Item # WW-22999-41
Eliminate multiple separate racks in your lab – here is an ideal solution for your different sized tubes!
  • Compact, modular design fits under lab hoods and preserves your valuable bench space
  • Polypropylene construction offers good chemical resistivity along with autoclavability
  • Interchangeable, color-coded inserts can also be used as stand-alone racks
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPipettingWorkStationwithoutbrackets(2/pk)
Key Features
  • Compact, modular design fits under lab hoods and preserves your valuable bench space
  • Polypropylene construction offers good chemical resistivity along with autoclavability
  • Interchangeable, color-coded inserts can also be used as stand-alone racks
More About this Item
This versatile pipetting workstation features a small footprint 25.4 x 12.7 x 7.6 cm (10" x 5" x 3") and its modular design allows you to create a system based on your individual requirements. The tiered rack design permits the simultaneous viewing of all tubes. Each rack accommodates up to six tube inserts or basins.
Color-coded polypropylene inserts accept tubes with sizes ranging from 0.5 to 50 mL. Easy-release tabs allow you to quickly change out the inserts to customize the rack to your applications. Each tube cavity is numbered for quick identification. Use the pipetting basin insert to hold up to 25 mL of reagent or solution. Use the bracket to connect two stations and have up to six tiers.