Cole-Parmer Essentials Inoculating Needles, 3", 25 Gauge; 12/Pk
Representative image only.
Cole-Parmer Essentials Item # WW-04396-49
Nichrome wire composition cools quickly and is less expensive than platinum
  • Select, inoculate, or transfer microbiological samples
PRODUCT OPTIONS
₹1,600.00INR / Pkg of 12
SPECS & DESCRIPTION
Specifications & Description
  • B and S Wire Gauge25
  • DescriptionInoculating Needles, 3\", 25 Gauge; 12/Pk
Key Features
  • Select, inoculate, or transfer microbiological samples
More About this Item
Needles are made from high quality nichrome wire that cools rapidly after sterilization. Available with or without handle and straight or angled needle.
Handles are not included with these needles. Brass handles are sold separately as part numbers 04396-40 and -41.