Cole-Parmer Essentials Inoculating Needle, Micro, Clear; 5/Pk
Representative image only.
Cole-Parmer Essentials Item # WW-04396-50
Nichrome wire composition cools quickly and is less expensive than platinum
  • Select, inoculate, or transfer microbiological samples
PRODUCT OPTIONS
₹8,700.00INR / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • ColorClear
  • DescriptionInoculating Needle, Micro, Clear; 5/Pk
Key Features
  • Select, inoculate, or transfer microbiological samples
More About this Item
Needles are made from high quality nichrome wire that cools rapidly after sterilization. Available with or without handle and straight or angled needle.
Attached to a light-weight aluminum handle, insulated to protect the hand during use.