Argos Technologies DRNAse Free™ Reagent Spray Bottle Surface Decontaminant; 720 mL
Representative image only.
Argos Technologies Item # WW-04397-24
Instantly eliminate DNase and RNase from labware and surfaces
  • Works on glass, plastic, stainless steel, and benchtops
  • Replace carcinogenic chemicals, like DEPC
  • Phosphate free and biodegradable
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)720
  • DescriptionReagent 720 mL Spray Bottle Surface Decontaminant
Key Features
  • Works on glass, plastic, stainless steel, and benchtops
  • Replace carcinogenic chemicals, like DEPC
  • Phosphate free and biodegradable
More About this Item
The DRNase Free reagent surface decontaminant is ideal for items and surfaces used in PCR that cannot be autoclaved. Just spray and rinse. This nontoxic formula doesn’t leave a residue, so it won’t interfere with gel polymerization or enzymatic reactions.
Certificate of Analysis
Enter lot number
or click to view a Sample C of A