Cole-Parmer Essentials Inoculating Loop and Needles, Sterile, Polystyrene, 10 µL, Individually Wrapped; 250/Cs
Representative image only.
Cole-Parmer Essentials Item # WW-06231-08
Sterile inoculating loops with needles save time and eliminate cross-contamination when transferring solid and liquid media
  • Loop and needle combination for dual function
  • Use straight from the pack without sterilizing
  • Flexible and disposable
  • Treated, hydrophilic surface
PRODUCT OPTIONS
₹4,600.00INR / Case of 250
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Overall Length (mm)228
  • Overall Length (in)8 31/32
  • Contained Volume (µL)10
  • ColorYellow
  • MaterialPolystyrene
  • DescriptionInoculating Loop and Needles, Sterile, Polystyrene, 10 µL, Individually Wrapped; 250/Cs
Sterile
Key Features
  • Loop and needle combination for dual function
  • Use straight from the pack without sterilizing
  • Flexible and disposable
  • Treated, hydrophilic surface
More About this Item
A loop on one end and needle on the other make these disposable inoculating lab tools suitable for both solid and liquid media. They are vacuum-gas plasma surface treated to allow culture medium to attach. The long, flexible stem of these inoculating loops can be bent allowing access to narrow or small-volume tubes and dishes. Color coding identifies inoculation volume easily.
Manufactured under ISO 13485 and ISO 9001 quality management systems in an ISO Class 8 (formerly Class 100,000) cleanroom environment, these radiation sterilized (SAL 10-6) inoculating loops are available in individual peel-to-open packs, or in resealable packs of 10 or 25. Every package is printed with a lot number for quality traceability. DNase/RNase free and nonpyrogenic.