Cole-Parmer Essentials Calibrated Inoculating Loops, 1 uL, 1.45 mm ID; 5/Pk
Representative image only.
Cole-Parmer Essentials Item # WW-04396-36
Nichrome wire composition cools quickly and is less expensive than platinum
  • Select, inoculate, or transfer microbiological samples
PRODUCT OPTIONS
₹29,700.00INR / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Loop Inside Diameter (mm)1.45
  • B and S Wire Gauge26
  • Contained Volume (µL)1
  • DescriptionCalibrated Inoculating Loops, 1 uL, 1.45 mm ID; 5/Pk
Key Features
  • Select, inoculate, or transfer microbiological samples
More About this Item
Argos brand Inoculating Loops are made from high quality twisted nichrome wire. Loops have been designed to slide along agar surfaces without cutting or tearing. Argos Inoculating Loops cool rapidly and are less expensive than platinum.
Please note: Loops are supplied without handles and are available calibrated or non-calibrated. Brass loop handles are sold separately as models 04396-40 and -41.
Calibrated Loops are supplied with calibration certificates based on ASM/APHA Evans Blue Dye test protocols.