Cole-Parmer Essentials Bulb Pipette Filler, Up to 50 mL, Red
Representative image only.
Cole-Parmer Essentials Item # WW-04395-02
Thicker rubber walls offer greater pipetting control
  • Manufactured for the safe and reliable aspiration and delivery of liquids
  • Ideal for use with serological pipettes up to 50mL
  • Constructed of durable red rubber
Often bought with…
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • AutoclavableYes
  • Max Volume (mL)50
  • Body MaterialRubber
  • Wetted MatterialRubber and Stainless Steel
  • Color CodeRed
  • Compatible withSerological pipettes up to 50-mL 
  • DescriptionBulb Pipette Filler, Up to 50 mL, Red
Key Features
  • Manufactured for the safe and reliable aspiration and delivery of liquids
  • Ideal for use with serological pipettes up to 50mL
  • Constructed of durable red rubber
More About this Item
Natural rubber pipette filler is ideal for serological pipettes with 50-mL capacity. Special pinch and release valve system with a chemical-resistant stainless steel ball bearing allows for easy one-hand operation. Available in a variety of color choices.