Cole-Parmer Essentials Inoculating Loop and Needle Holder, 20.32 cm (8 in), Brass
Representative image only.
Cole-Parmer Essentials Item # WW-04396-41
Brass holder safely and securely holds inoculating loops or needles
  • Helps prevent direct contact with sample
  • Insulated to protect hand during use
PRODUCT OPTIONS
₹5,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (mm)203.2
  • Overall Length (in)8
  • Handle TypeBrass
  • DescriptionInoculating Loop and Needle Holder, 20.32 cm (8 in), Brass
Key Features
  • Helps prevent direct contact with sample
  • Insulated to protect hand during use
More About this Item
These reusable brass handles securely hold inoculating loops and needles allowing you to select, inoculate or transfer microbiological samples during the streaking process. They are insulated to protect the hand during sterilization. Loops and needles are sold separately.