Applikon Fermentation Reagent Addition Bottle, 5.0 L
Representative image only.
Applikon Mfr # Z811301912 – Item # WW-01919-92
  • 1 year warranty
PRODUCT OPTIONS
₹241,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Liters)5
  • DescriptionFermentation Reagent Addition Bottle, 5.0 L
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Reagent Addition Bottles come in two sizes. Use small bottle for acids or bases; use large bottle to hold fresh media. Transfer pipes and filter included.
Autoclavable.