Applikon Fermentation/Bioreactor Addition Pipe for 10-mm Size Port
Representative image only.
Applikon Mfr # Z81324MT02 – Item # WW-01919-94
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFermentation/Bioreactor Addition Pipe for 10-mm Size Port
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Addition pipe is specially angled to allow reagent to drop gently on mediawon't harm cells growing near top of vessel.