Applikon Blind Caps; port size: 18 mm (M18 x 1.5 mm thread)
Representative image only.
Applikon Mfr # Z813010002 – Item # WW-01919-13
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • Connections18 mm port
  • DescriptionBlind Caps; port size: 18 mm (M18 x 1.5 mm thread)
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Blind Caps cover unused ports. Come complete with O-rings.