Applikon BioBundle Z010015420 Dissolved Oxygen Sensor for Fermentation System, 154 mm

Representative image only.
BioBundle Mfr # Z010015420 – Item # WW-29207-34
These sterilizable sensors offer guaranteed low drift
  • Fully electropolished surface (Ra < 0.8 µm) optimizes cleaning
  • Shaft diameter is 12 mm

SPECS & DESCRIPTION

Specifications & Description

  • DescriptionDissolved Oxygen Sensor for Fermentation System, 154 mm

Key Features

  • Fully electropolished surface (Ra < 0.8 µm) optimizes cleaning
  • Shaft diameter is 12 mm

More About this Item

All DO sensors are delivered factory tested with membrane module in place. All sensors can be mounted into virtually any brand of autoclavable or steam-in-place bioreactor.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat