Ansell Microflex® V29 Blue Disposable Vinyl Glove, Size XL (9.5-10); 100/Box
Representative image only.
Ansell Mfr # 163312 BOX – Item # WW-81610-55
Cost-effective protection for light duty tasks
  • Free from natural rubber latex
  • Produced without use of DEHP
  • Ambidextrous
PRODUCT OPTIONS
₹2,200.00INR / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialVinyl
  • Thickness (mil)2.8
  • SizeX-Large
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2600
  • SterileNo
  • PowderedNo
  • DescriptionBlue Disposable Vinyl Glove, Size XL (9.5-10); 100/Box
Key Features
  • Free from natural rubber latex
  • Produced without use of DEHP
  • Ambidextrous
More About this Item
These disposable blue vinyl gloves provide outstanding elasticity and tensile strength, ideal for food preparation, light-duty cleaning, and general maintenance. The gloves offer a smooth finish, giving you a more natural fit. Powder-free to reduce skin irritation. Disposable—for single use only.