Ansell Microflex® Dura Flock® DFK-608-S Flock-Lined Disposable Nitrile Gloves, Small; 500/CS (10 boxes of 50 gloves)
Representative image only.
Ansell Mfr # 162598 – Item # WW-81620-65
Moisture-absorbing flock lining keeps your hands dry and comfortable
  • Thick design resists punctures and tears
  • Durable nitrile for reliable splash protection
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)8
  • SizeSmall (6.5-7.0)
  • Length (in)10 1/2
  • Length (cm)27
  • ColorDark Green
  • LiningFlock-lined
  • GripTextured
  • SterileNo
  • PowderedPowder free
  • DescriptionFlock-Lined Disposable Nitrile Gloves, Small; 500/CS
Key Features
  • Thick design resists punctures and tears
  • Durable nitrile for reliable splash protection
More About this Item
These disposable, 100% nitrile gloves feature a thin layer of fine cotton that absorbs moisture, keeping your hands comfortable and reducing hand fatigue even for extended periods. Glove thickness of 8 mil increases durability and offers superior protection. Gloves are fully textured for a firm, reliable grip. Powder free. Ambidextrous.