Ansell HyFlex® 11-100 Durable Work Gloves with Antimicrobial Material, Size 10; 12 Pair/Bag
Representative image only.
Ansell Mfr # 103344 – Item # WW-81620-96
Lifelong antimicrobial protection
  • Touchscreen compatible
  • Comfort fit yarn with nitrile foam coating offers all-day comfort
  • Protection from bacterial-related odor, stain, and fabric degradation
  • Self-Sanitizing 360⁰ antimicrobial protection of both liner and coating
  • Compliant with EU Biocides Regulation 528/2012
PRODUCT OPTIONS
₹21,800.00INR / Bag of 12
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Size10
  • Length (in)10 25/32
  • Length (cm)27.4
  • ColorGrey
  • LiningConductive fiber, nylon, Ionic+ active antimicrobial liner
  • CuffKnitwrist
  • SterileNo
  • DescriptionDurable Work Gloves with Antimicrobial and Touch Screen Material, Size 10; 12 Pair/Bag
Key Features
  • Touchscreen compatible
  • Comfort fit yarn with nitrile foam coating offers all-day comfort
  • Protection from bacterial-related odor, stain, and fabric degradation
  • Self-Sanitizing 360⁰ antimicrobial protection of both liner and coating
  • Compliant with EU Biocides Regulation 528/2012
More About this Item
These industrial gloves feature antimicrobial protection and touchscreen compatible with a wide range of devices. Designed with Inoic+™, an EPA-registered antimicrobial technology, the glove coating does not inhibit antimicrobial action. Ionic+ is permanent and lasts the life of the product. The light-weight breathable glove design provides you all day comfort with high dexterity.