Ansell AlphaTec® 58-128 Light Duty Durable Nitrile Gloves, Size 9; 12 Pair/Bag
Representative image only.
Ansell Mfr # 121620 – Item # WW-81620-87
Special coating enhances dexterity, grip, and comfort
  • First ergonomically certified chemical glove
  • Delivers high-performance musculoskeletal support
  • Reduces risk of ergonomic injury
  • 12 pairs (24 gloves) per polybag
PRODUCT OPTIONS
₹13,600.00INR / Bag of 12
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Size9
  • Length (in)11
  • Length (cm)28
  • ColorGray and Green
  • GripAnsell Grip™ Technology
  • CuffStraight
  • SterileNo
  • DescriptionLight Duty Durable Nitrile Gloves, Size 9; 12 Pair/Bag
Key Features
  • First ergonomically certified chemical glove
  • Delivers high-performance musculoskeletal support
  • Reduces risk of ergonomic injury
  • 12 pairs (24 gloves) per polybag
More About this Item
These ergonomically certified chemical gloves offer musculoskeletal support and reduce the risk of injury caused by prolonged or repetitive tasks. A thin nitrile barrier protects you from oils and other irritant fluids, while a special grip technology makes handling wet or oily parts safe and easy. The gloves conform to the shape of your hand enabling natural movement.