Ansell TouchNTuff® 92-675 Nitrile Gloves with Textured Fingertips, Size L (8.5-9); Case of 1000 (10 boxes of 100 gloves)
Representative image only.
Ansell Mfr # 105083 – Item # WW-81602-78
Robust gloves offer excellent chemical splash protection
  • Nitrile is a durable and soft material for strength and comfort
  • Get a secure grip with textured fingertips
  • Complies with FDA food handling requirements
  • Large size
PRODUCT OPTIONS
₹50,200.00INR / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)4.3
  • SizeLarge
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • GripTextured
  • CuffBeaded
  • ComplianceCategory III, EN ISO 374-1:2016, EN 374:2003, EN 420:2003 +A1:2009, EN ISO 374-5:2016, FDA21 CFR 177-2600, ISO 9001
  • PowderedNo
  • DescriptionNitrile Gloves with Textured Fingertips, Size L (8.5-9.); Case of 1000 (10 boxes of 100 gloves)
Key Features
  • Nitrile is a durable and soft material for strength and comfort
  • Get a secure grip with textured fingertips
  • Complies with FDA food handling requirements
  • Large size
More About this Item
TouchNTuff disposable powder-free nitrile gloves are appropriate for transferring liquids or solids, sample taking/processing, handling of incoming goods, maintenance, and food processing. Textured fingertips enhance wet or dry grip. Ambidextrous with beaded cuffs. Blue color.
Compliance: FDA 21 CFR Part 177.2600, ISO 9001