Ansell 224 192243 Flock-Lined, Neoprene-Over-Latex Gloves, size 8, 12 pair/pk
Representative image only.
Ansell Mfr # 224 192243 – Item # WW-81601-30
Two-tone color indicates glove wear over time
  • Protects against most chemicals
  • Lining absorbs perspiration for increased comfort
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)28
  • MaterialNeoprene
  • Size8
  • Length (in)13
  • Length (cm)33
  • ColorBlue and Yellow
  • LiningCotton
  • GripDiamond-embossed
  • SterileNo
  • DescriptionFlock-Lined, Neoprene-Over-Latex Gloves, size 8, 12 pair/pk
Key Features
  • Protects against most chemicals
  • Lining absorbs perspiration for increased comfort
More About this Item
Gloves cost less than neoprene but still provide excellent protection against most chemicals. The 100% cotton flock lining serves as an extra layer of protection and absorbs perspiration for increased comfort. Recessed diamond pattern grip.