Analytik Jena 230 VAC Imaging System
Representative image only.
Analytik Jena Mfr # 97-0109-02 – Item # 9770135
Low profile viewing cabinet with overhead UV
  • DigiCam - digital color camera with 24 megapixel resolution
  • Lightweight cabinet
  • 254 nm and 365 nm overhead UV illumination
  • Capture software enables live preview of images
  • 1 year warranty
₹1,608,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 year
1 Year Warranty
Key Features
  • DigiCam - digital color camera with 24 megapixel resolution
  • Lightweight cabinet
  • 254 nm and 365 nm overhead UV illumination
  • Capture software enables live preview of images
  • 1 year warranty
More About this Item
The UVP ChromaDoc-It System connects to your computer for easy documentation of thin layer chromatography. The lightweight viewing cabinet is perfect for TLC applications with overhead 15-watt 254 nm and 365 nm UV illumination. The high resolution Digi Camera is controlled by Analytik Jena's exclusive capture software. The software enables live preview of images, user defined default settings and templates, annotations, use of filters, and report generation.